Order Entry
Austria
Orders LinkContactUsLinkComponent
Poster - Digestive System

Poster - Digestive System

Wall chart: Digestive system, 840×1180 mm
Catalog # 763-0162
Marketsource
Supplier:   3B Scientific
Poster - Digestive System
Catalog # 763-0162
Supplier:  3B Scientific
Marketsource
Poster - Digestive System
Poster - Digestive System

Wall chart: Digestive system, 840×1180 mm

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents

Plank mounted on roll. 84×118 cm.

Specifications
  • Description:
    Wall chart: Digestive system
  • W×H:
    840×1180 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.