Order Entry
Austria
Orders LinkContactUsLinkComponent
Heart

Heart

Classic heart in two parts, 190×120×120 mm
Catalog # 763-0116
Marketsource
Supplier:   3B Scientific
Heart
Catalog # 763-0116
Supplier:  3B Scientific
Marketsource
Heart
Heart

Classic heart in two parts, 190×120×120 mm

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents

Simple heart model in 2 parts. Atrium and chambers are clearly exposed. Dimensions 19×12×12 cm.

Specifications
  • Description:
    Classic heart in two parts
  • W×D×H:
    190×120×120 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.