ContactUsLinkComponent
Head with muscles, half
Head with muscles, half
  763-0100
undefined
Head with muscles, half
  763-0100
  • Description:
    Half head with muscles
  • Pk:
    1
The model shows all external structures such as blood vessels, nerves, etc. The median section shows all the structures of the head and neck. Dimensions: 22×18×46 cm.