Orders LinkContactUsLinkComponent
Leader, function models
Leader, function models
Marketsource
Supplier:   3B Scientific
Leader, function models
Supplier:  3B Scientific
Marketsource
Certificates
Leader, function models
Leader, function models
3 Options Available Below
Product Details & Documents
Shoulder joint: Life-size plastic moldings of the upper part of the humerus; shoulder blades and collarbones with ligaments. Movements that can be studied: the arm is lifted (abduction), moved forwards and backwards (anteversion or retroversion) and rotation inwards and outwards. On stand.
Hip joint: Natural casting. Consists of the hip bone and the upper part of the femur with ligaments. On stand. Movements that can be shown are anteversion, retroversion and abduction.
Elbow joint: Natural casting. Consists of upper arm and forearm with ligaments. On stand. Functions that can be shown are bending and stretching of the upper and forearm as well as the rotational movements of the arm.
Product Family Options
Product Information
  • Availability
    Price
  • 763-0086
    Shoulder joint, function model
    Shoulder joint, function model
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 190,00 €
    190,00 €
    1 * 1 ST
       190,00 €
    Specifications:
    Marketsource
    Marketsource
  • 763-0087
    Hip joint, function model
    Hip joint, function model
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 190,00 €
    190,00 €
    1 * 1 ST
       190,00 €
    Specifications:
    Marketsource
    Marketsource
  • 763-0089
    Elbow joint, function model
    Elbow joint, function model
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 190,00 €
    190,00 €
    1 * 1 ST
       190,00 €
    Specifications:
    Marketsource
    Marketsource