Order Entry
Austria
Orders LinkContactUsLinkComponent
Leaf structure model

Leaf structure model

Leaf structure model, Fagus silvatica, 1 kg, 300×300×120 mm
Catalog # 763-0137
Marketsource
Supplier:   3B Scientific
Leaf structure model
Catalog # 763-0137
Supplier:  3B Scientific
Marketsource
Leaf structure model
Leaf structure model

Leaf structure model, Fagus silvatica, 1 kg, 300×300×120 mm

Price
553,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Fagus silvatica. Three dimensional model of an beech tree leaf at 1500X magnification.

Shows different cell types and stomata.

Specifications
  • Weight:
    1 kg
  • Description:
    Leaf structure model, Fagus silvatica
  • W×D×H:
    300×300×120 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.