Order Entry
Austria
Orders LinkContactUsLinkComponent
Buffer, sodium acetate buffer solution pH 5,2 (3 mol/L)

Buffer, sodium acetate buffer solution pH 5,2 (3 mol/L)

Sodium acetate buffer solution pH 5,2 (3 mol/l), Glass bottle, 100 mL
Catalog # ROCKMB-016
Buffer, sodium acetate buffer solution pH 5,2 (3 mol/L)
Catalog # ROCKMB-016
CAS Number:  
Buffer, sodium acetate buffer solution pH 5,2 (3 mol/L)
Buffer, sodium acetate buffer solution pH 5,2 (3 mol/L)

Sodium acetate buffer solution pH 5,2 (3 mol/l), Glass bottle, 100 mL

CAS Number:  
Price
184,00 €
Log in for availability
1 * 100 mL
Product Details & Documents

This product was aseptically filtered through a Millipore 0,22 µm filter into clean, pre-sterilised containers. The product was tested on trypticase soy agar for 24 hours, 48 hours and 72 hours and was found to be negative for bacteria.

Caution: This product is for research use only and is not intended for therapeutic or diagnostic applications.

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Description:
    Sodium acetate buffer solution pH 5,2 (3 mol/l)

Specifications