Order Entry
Austria
Orders LinkContactUsLinkComponent
Buffer, sodium acetate buffer solution pH 5,2 (3 mol/l)

Buffer, sodium acetate buffer solution pH 5,2 (3 mol/l)

Sodium acetate buffer solution pH 5,2 (3 mol/l), Glass bottle, 100 mL
Catalog # R010
Supplier:   G-Biosciences
Buffer, sodium acetate buffer solution pH 5,2 (3 mol/l)
Catalog # R010
Supplier:  G-Biosciences
CAS Number:  
Buffer, sodium acetate buffer solution pH 5,2 (3 mol/l)

Sodium acetate buffer solution pH 5,2 (3 mol/l), Glass bottle, 100 mL

CAS Number:  
Price
45,80 €
Log in for availability
1 * 100 mL
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Description:
    Sodium acetate buffer solution pH 5,2 (3 mol/l)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.