Digital pH Sensors, Memosens CPS11E
Supplier: Endress+Hauser
New Product
Certificates
About this item
Digital pH sensor for standard applications with contactless signal transmission for safe processes.
- Robust electrode: Special design prevents poisoning, with a dirt-repellent PTFE junction guarding against medium contamination
- Durable materials: Process glass handles full pH range, tolerates up to 17 bar pressure (Optional F-glass suits hydrofluoric acid, like in semiconductor baths)
- Fast sensor replacement reduces process downtime, extends sensor life and minimizes operating costs
- Increased reliability in low conductivity applications: An improved optional salt ring ensures reliable measurements in scenarios such as boiler feed water
- Memosens 2.0 technology ensures data storage for better trend analysis, predictive maintenance, and advanced IIoT functions
The Memosens CPS11E, a digital pH sensor, guarantees process stability and provides reliable measured values, even in harsh pH environments or in risk zones (depending on the model). Thanks to Memosens 2.0 digital technology, this sensor is moisture resistant and supports laboratory calibration, which increases process safety and reduces operational complexity. With extended storage of calibration and process data, it provides a solid basis for predictive maintenance. This sensor is designed for durability and minimal maintenance, ensuring a long service life.
Ordering information: For optional items or more information, contact your Avantor representative.
Delivery information: Digital pH sensor Memosens CPS11E.
Specifications
- Catalog No:
- ENDRCPS11E-10Q9/0
- ENDRCPS11E-10K6/0
- ENDRCPS11E-10P3/0
- ENDRCPS11E-1082/0
- ENDRCPS11E-1035/0
- ENDRCPS11E-12H3/0
- ENDRCPS11E-1014/0
- Sensor type:
- NTC (RTD type)
- NTC (RTD type)
- NTC (RTD type)
- NTC (RTD type)
- NTC (RTD type)
- NTC (RTD type)
- NTC (RTD type)
- Shaft material:
- Glass
- Glass
- Glass
- Glass
- Glass
- Glass
- Glass