Order Entry
Austria
ContactUsLinkComponent
Organic reference standard, Cumene 2,000 µg/ml in methanol
Organic reference standard, Cumene 2,000 µg/ml in methanol
Supplier:  Restek
Certificates
Organic reference standard, Cumene 2,000 µg/ml in methanol
Supplier:  Restek

Specifications

  • CAS No.:
    98-82-8
  • Formula:
    C₉H₁₂ 
  • Description:
    Cumene

Specifications

Frequently Bought Together