Order Entry
Austria
ContactUsLinkComponent
Buffer, TAE solution 50X concentrate (TRIS-acetate-EDTA buffer)
Buffer, TAE solution 50X concentrate (TRIS-acetate-EDTA buffer)
Catalog # ROCKMB-020
CAS Number:  
undefined
Buffer, TAE solution 50X concentrate (TRIS-acetate-EDTA buffer)
Catalog # ROCKMB-020
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Description:
    TAE buffer solution 50X concentrate (TRIS-acetate-EDTA buffer)

Specifications

About this item