Order Entry
Austria
ContactUsLinkComponent
 
Lecithin (from soy bean), granular
 
undefined
Lecithin (from soy bean), granular
 

 

  • Appearance:
    Yellow to tan Granules
  • Infrared spectrum:
    Conforms
  • Assay:
    ≥97 % (Phospholipids) as Acetone insolubles
  • Water:
    ≤1.1 % (K.F.)
  • Insoluble matter:
    ≤0.02 % (in hexane)
  • Acid value:
    ≤35 mg KOH/g
  • Peroxide value:
    ≤4 meq O2/kg
  • UV:
    at 280 nm A: ≤0.1
  • Residue after ign. (sulfated):
    ≤0.3 %
  • Specific optical rotation:
    +102° to +106° (20°C, 589 nm) (10% in water)
  • Heavy metals (as Pb):
    ≤10 ppm

 

 

Frequently Bought Together