Lecithin from egg yolk
Supplier: PanReac AppliChem
Certificates
Certificates
About this item
CAS Number:93685-90-6
:
MDL Number:MFCD00131449
EINECS:297-639-2
Frequently Bought Together
Spare Parts for VWR® Hitachi HPLC Instruments
Supplier: VWR Collection
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Test tube racks, Unwire™, Half-Racks, Nalgene®
Supplier: Thermo Fisher Scientific
VWR®, Nitrile Gloves
Supplier: VWR Collection
Tubing clamps, stainless steel
Supplier: USBECK Laborgeräte
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Dehydrated culture media
Supplier: Merck
VWR®, Spreaders, L-shaped, Disposable
Supplier: VWR Collection
Polysorbate 80, GPR RECTAPUR®
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
CASEIN FROM BOVINE MILK SODIUM 1 * 1 kg
Supplier: Merck
Casein, enzymatic hydrolysate
Supplier: Apollo Scientific
VWR® Culture Media, Ready To Use, In Plates
Supplier: VWR Chemicals