Order Entry
Austria
Orders LinkContactUsLinkComponent
Statine ≥95%

Statine ≥95%

Plastic bottle, 100 mg
Catalog # AATB5109
Irritant
Supplier:   AAT Bioquest
Statine ≥95%
Catalog # AATB5109
Supplier:  AAT Bioquest
CAS Number:  49642-07-1
Statine ≥95%
Statine ≥95%

Plastic bottle, 100 mg

CAS Number:  49642-07-1
Price
313,00 €
Log in for availability
1 * 100 mg
Product Details & Documents

(3S,4S)-Statine is a novel amino acid of pepstatin, a low molecular weight inhibitor of acid proteases.

  • CAS Number:49642-07-1
  • Formula:C₈H₁₇NO₃
  • MDL Number:MFCD00037277
  • Molecular Weight:175.228 g/mol
  • ADR:5,II
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.