Order Entry
Austria
ContactUsLinkComponent
7-Amino-4-methyl-3-coumarinylacetic acid (AMCA-H) ≥95%
 
7-Amino-4-methyl-3-coumarinylacetic acid (AMCA-H) ≥95%
  AATB501
 :  
undefined
 
7-Amino-4-methyl-3-coumarinylacetic acid (AMCA-H) ≥95%
  AATB501
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    25 mg
  • Seq:
    2315

 

 

 :
 :233.22 g/mol
 :5,II
 :GHS07
 :106562-32-7
 :C₁₂H₁₁NO₄
 :MFCD00081051