Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
DBCO-NHS ester
DBCO-NHS ester for biomolecular labeling
DBCO-NHS ester for biomolecular labeling
AATB4529
:
AAT BIOQUEST
:
DBCO-NHS ester for biomolecular labeling
AATB4529
:
AAT BIOQUEST
:
Pack type:
Plastic bottle
Pk:
10 mg
Seq:
2275
:
:
402.41 g/mol
:
5,II
:
GHS07
:
1353016-71-3
:
C₂₃H₁₈N₂O₅
:
MFCD24386367