0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Austria
ContactUsLinkComponent
iFluor®
iFluor® 750, Styramide *Superior Replacement for Alexa Fluor 750 tyramide*
iFluor® 750, Styramide *Superior Replacement for Alexa Fluor 750 tyramide*
iFluor® 750, Styramide *Superior Replacement for Alexa Fluor 750 tyramide*
Catalog # AATB45065
Supplier:
AAT BIOQUEST
CAS Number:
Expand image
iFluor® 750, Styramide *Superior Replacement for Alexa Fluor 750 tyramide*
Catalog # AATB45065
Supplier:
AAT BIOQUEST
CAS Number:
Specifications
Pk:
100
Seq:
0100
Specifications
About this item
Product Details
:
Product Details