Order Entry
Austria
ContactUsLinkComponent
5(6)-Carboxyrhodamine 6G cadaverine ≥95%
5(6)-Carboxyrhodamine 6G cadaverine ≥95%
Catalog # AATB343
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
5(6)-Carboxyrhodamine 6G cadaverine ≥95%
Catalog # AATB343
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :770.72 g/mol