Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
APC-Cy7 Tandem
APC-Cy7 Tandem, ReadiUse™ fluorescent probe, preactivated
APC-Cy7 Tandem, ReadiUse™ fluorescent probe, preactivated
APC-Cy7 Tandem, ReadiUse™ fluorescent probe, preactivated
Catalog # AATB2587
Supplier:
AAT BIOQUEST
CAS Number:
Expand image
APC-Cy7 Tandem, ReadiUse™ fluorescent probe, preactivated
Catalog # AATB2587
Supplier:
AAT BIOQUEST
CAS Number:
Specifications
Pack type:
Plastic bottle
Pk:
1 mg
Seq:
2070
Specifications
About this item
Product Details
:
:
105000 g/mol
ADR:
5,II
:
GHS07
Product Details