Order Entry
Austria
ContactUsLinkComponent
Tide Quencher™ 2 phosphoramidite, Tide Quencher™ fluorescent probe
Irritant
Tide Quencher™ 2 phosphoramidite, Tide Quencher™ fluorescent probe
Catalog # AATB2208
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Irritant
Tide Quencher™ 2 phosphoramidite, Tide Quencher™ fluorescent probe
Catalog # AATB2208
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 µmol
  • Seq:
    9140

Specifications

About this item

 :
 :681.87 g/mol
ADR:5,II
 :GHS07