Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
DABCYL-DBCO
DABCYL-DBCO FRET acceptor precursor
DABCYL-DBCO FRET acceptor precursor
AATB2010
:
AAT BIOQUEST
:
DABCYL-DBCO FRET acceptor precursor
AATB2010
:
AAT BIOQUEST
:
Pack type:
Plastic bottle
Pk:
5 mg
Seq:
2210
:
:
5,II
:
527.63 g/mol
:
GHS07