Order Entry
Austria
ContactUsLinkComponent
Probenecid ≥95% cell culture tested
Irritant
Probenecid ≥95% cell culture tested
Catalog # AATB20060
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Irritant
Probenecid ≥95% cell culture tested
Catalog # AATB20060
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    720 mg
  • Seq:
    2797

Specifications

About this item

 :
 :285.4 g/mol
UN:3249
ADR:5,II
 :GHS07
CAS Number:57-66-9
Formula:C₁₃H₁₉NO₄S
MDL Number:MFCD00038402