
Thiazole orange ≥90%
Catalog # AATB17518
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:476.61 g/mol
ADR:5,II
:GHS07
CAS Number:107091-89-4
Formula:C₂₆H₂₄N₂O₃S₂
MDL Number:MFCD00192499