0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Austria
ContactUsLinkComponent
Cyanine-5- dUTP [Cy5-dUTP]
Cyanine-5- dUTP [Cy5-dUTP] 1 mM in TRIS buffer (pH 7.5)
Cyanine-5- dUTP [Cy5-dUTP] 1 mM in TRIS buffer (pH 7.5)
Cyanine-5- dUTP [Cy5-dUTP] 1 mM in TRIS buffer (pH 7.5)
Catalog # AATB17026
Supplier:
AAT BIOQUEST
CAS Number:
Expand image
Cyanine-5- dUTP [Cy5-dUTP] 1 mM in TRIS buffer (pH 7.5)
Catalog # AATB17026
Supplier:
AAT BIOQUEST
CAS Number:
Specifications
Pack type:
Plastic bottle
Pk:
25 Nmo
Seq:
0311
Specifications
About this item
Product Details
:
ADR:
5,II
:
1176.04 g/mol
:
GHS07
Product Details