Order Entry
Austria
ContactUsLinkComponent
 
FCB [Fluorescein di-beta-D-cellobioside]
Catalog # AATB14025
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
FCB [Fluorescein di-beta-D-cellobioside]
Catalog # AATB14025
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :980.87 g/mol
ADR:5,II
 :GHS07