0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Austria
ContactUsLinkComponent
Z-IETD-FMK
Z-IETD-FMK
Z-IETD-FMK
Z-IETD-FMK
AATB13305
:
AAT BIOQUEST
:
Z-IETD-FMK
AATB13305
:
AAT BIOQUEST
:
Pack type:
Plastic bottle
Pk:
1 mg
Seq:
2070
:
:
654.68 g/mol
:
5,II
:
GHS07
:
210344-98-2
:
C₃₀H₄₃FN₄O₁₁
:
MFCD03490491