Screen Quest™ Colorimetric ELISA cAMP Assay Kit
Supplier: AAT BIOQUEST
Certificates
About this item
Adenosine 3’, 5’ cyclic monophosphate (cAMP) is an important second messenger in intracellular signal transduction.
- No acetylation step
- Robust protocol
- Sensitive substrate for easy quantification
Caution: For research use only.
Specifications
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Conjugate ELISA:HRP
- Format:Pre-coated
- Host:Human
- Primary antibody reactivity:Human
- Target protein:cAMP
- Detection method:Colorimetric microplate reader
Specifications
Related Information
Frequently Bought Together