Native Streptomyces avidinii Streptavidin (from Streptomyces avidinii), APC (Allophycocyanin)
Catalog # AATB16902
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pack type:Vial
- Pk:100 µG
- Conjugation:APC (Allophycocyanin)
- Protein/peptide type:Native
- Source:Streptomyces avidinii
- Species:Streptomyces avidinii
- Protein synonyms:Avidin, Streptomyces avidinii
- Protein/peptide name:Streptavidin
Specifications
About this item
Streptavidin conjugates are widely used together with a conjugate of biotin for specific detection of a variety of proteins, protein motifs, nucleic acids and other molecules since streptavidin has a very high binding affinity for biotin.
- Extremely high biotin-binding affinity
- High quality conjugate
Caution: For research use only.