Order Entry
Austria
ContactUsLinkComponent
Native Bovine Bovine Serum Albumin (from Serum), Spectinomycin
Native Bovine Bovine Serum Albumin (from Serum), Spectinomycin
Catalog # AATB50038
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Native Bovine Bovine Serum Albumin (from Serum), Spectinomycin
Catalog # AATB50038
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    1 mg
  • Conjugation:
    Spectinomycin
  • Protein/peptide type:
    Native
  • Source:
    Serum
  • Species:
    Bovine
  • Protein synonyms:
    BSA
  • Protein/peptide name:
    Bovine Serum Albumin

Specifications

About this item

Spectinomycin-BSA conjugate is used for developing anti-spectinomycin antibodies that are typically used in an ELISA assay.

  • High immunogenicity
  • Useful for antibody development
  • ELISA assay building block
Caution: For research use only.