Synthetic Cephaelis ipecacuanha Cephaeline (from Cephaelis ipecacuanha), Bodi Fluor™ 488
Catalog # AATB32000
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pack type:Vial
- Pk:1 mg
- Conjugation:Bodi Fluor™ 488
- Protein/peptide type:Synthetic
- Source:Cephaelis ipecacuanha
- Species:Cephaelis ipecacuanha
- Environmentally Preferable:
- Protein synonyms:7',10,11-Trimethoxyemetan-6'-ol|Cepheline
- Protein/peptide name:Cephaeline
Specifications
About this item
Cephaeline is an alkaloid that is found in Cephaelis ipecacuanha and other plant species including Psychotria acuminate.
- Expectorant
- Multiple clinical applications
Caution: For research use only.