Order Entry
Austria
ContactUsLinkComponent
Native Bovine Bovine Serum Albumin (from Serum), Fentanyl
Native Bovine Bovine Serum Albumin (from Serum), Fentanyl
  AATB50565
 :  
undefined
Native Bovine Bovine Serum Albumin (from Serum), Fentanyl
  AATB50565
 :  

 

  • Pack type:
    Vial
  • Pk:
    1 mg
  • Conjugation:
    Fentanyl
  • Protein/peptide type:
    Native
  • Source:
    Serum
  • Species:
    Bovine
  • Protein synonyms:
    BSA
  • Protein/peptide name:
    Bovine Serum Albumin

 

 

Fentanyl-BSA conjugate is widely used for screening fentanyl antibodies and developing fentanyl assays and tests.

  • High immunogenicity
  • Useful for antibody development
  • Fentanyl test building block
 : For research use only.