Orders LinkContactUsLinkComponent
Cell Meter™ IX830 Fixable Viability Dye
Cell Meter™ IX830 Fixable Viability Dye
Cell Meter™ IX830 fixable cell stain, 200 Tests, 200 Tests
Catalog # AATB22529
Supplier:   AAT Bioquest
Cell Meter™ IX830 Fixable Viability Dye
Catalog # AATB22529
Supplier:  AAT Bioquest
CAS Number:  
Cell Meter™ IX830 Fixable Viability Dye
Cell Meter™ IX830 Fixable Viability Dye
Cell Meter™ IX830 fixable cell stain, 200 Tests, 200 Tests
CAS Number:  
Price
299,00 €
Log in for availability
1 * 200 Tests
Product Details & Documents
Discrimination and exclusion of dead cells from live cells allows cleaner separation and identification of cell populations.
  • Cell-impermeable
  • Amine- and thiol-reactive
  • High stability
  • Live/dead cell labeling dye
  • Fixable
Caution: For research use only.
Specifications
Specifications
  • Pk:
    200 Tests
  • Description:
    Cell Meter™ IX830 fixable cell stain
  • Tests per kit:
    200 Tests
Specifications