hBSA *Optimized for Maximum Immunization Response* (10 mg)
Catalog # AATB5601
Supplier: AAT BIOQUEST
Specifications
- Serum format:Lyophilised
- Sterility:Non sterile
- Description:BSA cationized with hexamethylenediamine
- Storage temperature:<–20 °C
- Purification:97%, via fractionation
- Pack type:Bottle
- Pk:10 mg
Specifications
About this item
hBSA is the bovine serum albumin modified with hexamethylenediamine to make the proteins bear more amine groups (thus more positive charges at neutral pH).
- Lyophilized powder
- Excellent quality control
- Stronger immune response
- 60000 molecular weight
Caution: For research use only.