Order Entry
Austria
ContactUsLinkComponent
FluoroQuest™ Mounting Medium with DAPI (50 ml)
FluoroQuest™ Mounting Medium with DAPI (50 ml)
Catalog # AATB20004
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
FluoroQuest™ Mounting Medium with DAPI (50 ml)
Catalog # AATB20004
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    50 mL
  • Description:
    FluoroQuest™ with DAPI

Specifications

About this item

FluoroQuest™ with DAPI is an aqueous mounting medium for preserving fluorescence of tissue and cell smears.

  • Contains DNA/RNA counterstain DAPI
  • Improves and sustains fluorescence
  • Stable for extended storage