Order Entry
Austria
ContactUsLinkComponent
MitoLite™ Deep Red FX660
MitoLite™ Deep Red FX660
Catalog # AATB22678
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
MitoLite™ Deep Red FX660
Catalog # AATB22678
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    500 Tests
  • Description:
    MitoLite™ deep red
  • Purity:
    95%
  • Tests per kit:
    500 Tests

Specifications

About this item

MitoLite™ deep red is a cationic dye that selectively accumulates in mitochondria probably vial the mitochondrial membrane potential gradient.

  • Mitochondria-Specific Dye
  • Cell-Permeable
  • Robust
Caution: For research use only.