ContactUsLinkComponent
ddGTP [2',3'-Dideoxyguanosine-5'-triphosphate] *10 mM in ddH₂O* (1 µmole)
ddGTP [2',3'-Dideoxyguanosine-5'-triphosphate] *10 mM in ddH₂O* (1 µmole)
  AATB17210
 :  
undefined
ddGTP [2',3'-Dideoxyguanosine-5'-triphosphate] *10 mM in ddH₂O* (1 µmole)
  AATB17210
 :  
  • Pk:
    1 µmol
  • Description:
    ddGTP [2',3'-dideoxyguanosine-5'-triphosphate] *10 mm in ddH₂O*
Sanger sequencing, also known as the chain termination method, is a technique for DNA sequencing based upon the selective incorporation of chain-terminating dideoxynucleotides (ddNTPs) by DNA polymerase.
  • In-house quality control
  • Useful for sanger sequencing applications
 : For research use only.