Order Entry
Austria
ContactUsLinkComponent
ReadiLink™ Rapid iFluor® Antibody Labeling Kit "Microscale Optimized for Labeling 50 µg Antibody Per Reaction" (2 Labelings)
ReadiLink™ Rapid iFluor® Antibody Labeling Kit "Microscale Optimized for Labeling 50 µg Antibody Per Reaction" (2 Labelings)
Catalog # AATB1255
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
ReadiLink™ Rapid iFluor® Antibody Labeling Kit "Microscale Optimized for Labeling 50 µg Antibody Per Reaction" (2 Labelings)
Catalog # AATB1255
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    2 Tests
  • Description:
    ReadiLink™ rapid iFluor™ 488 antibody labelling kit 'Microscale optimized for labelling 50 µg antibody per reaction'

Specifications

About this item

Although FITC is still the most popular fluorescent labelling dye for preparing green fluorescent bioconjugates, there are certain limitations with FITC, such as severe photobleaching for microscope imaging and pH-sensitive fluorescence.

  • Contains all essential components
  • Minimal hands on time
  • Ready to use
  • Robust and simple antibody labelling protocol
  • No column purification required
Caution: For research use only.