Amplite® Rapid Fluorimetric Glutathione GSH/GSSG Ratio Assay Kit *Green Fluorescence*
Catalog # AATB10060
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pk:200 Tests
- Description:Amplite™ rapid fluorimetric glutathione GSH/GSSG ratio assay kit *green fluorescence*
- Tests per kit:200 Tests
Specifications
About this item
When cells are exposed to increased levels of oxidative stress, GSSG will accumulate and the ratio of GSH to GSSG will decrease.
- In-house QC testing
- Sensitive
- Suggested protocol available
Caution: For research use only.