ContactUsLinkComponent
Amplite® Colorimetric Beta-Galactosidase Assay Kit
Amplite® Colorimetric Beta-Galactosidase Assay Kit
Catalog # AATB12604
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Amplite® Colorimetric Beta-Galactosidase Assay Kit
Catalog # AATB12604
Supplier:  AAT BIOQUEST
CAS Number:  
Specifications
  • Pk:
    200 Tests
  • Description:
    Amplite™ colorimetric beta-galactosidase assay kit
Specifications
About this item
E. coli beta-galactosidase is a 464 kD tetramer.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Caution: For research use only.