ContactUsLinkComponent
Amplite® Fluorimetric NADP/NADPH Ratio Assay Kit *Red Fluorescence*
Amplite® Fluorimetric NADP/NADPH Ratio Assay Kit *Red Fluorescence*
Catalog # AATB15264
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Amplite® Fluorimetric NADP/NADPH Ratio Assay Kit *Red Fluorescence*
Catalog # AATB15264
Supplier:  AAT BIOQUEST
CAS Number:  
Specifications
  • Pk:
    250 Tests
  • Description:
    Amplite™ fluorimetric NADP/NADPH ratio assay kit *red fluorescence*
Specifications
About this item
Nicotinamide adenine dinucleotide (NAD⁺) and nicotinamide adenine dinucleotide phosphate (NADP⁺) are two important cofactors found in cells.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Caution: For research use only.