Order Entry
Austria
ContactUsLinkComponent
FluoroQuest™ Anti-Fading Kit II *Optimized for Plate Imaging* (1 Kits)
FluoroQuest™ Anti-Fading Kit II *Optimized for Plate Imaging* (1 Kits)
Catalog # AATB20003
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
FluoroQuest™ Anti-Fading Kit II *Optimized for Plate Imaging* (1 Kits)
Catalog # AATB20003
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    FluoroQuest™ anti-fading kit II *optimized for plate imaging*

Specifications

About this item

When exposed to excitation light, fluorescence intensity of dyes decreases due to their photooxidation or other photoreactions.

  • Preserves fluorescence
  • Prevents photobleaching
  • Ready to use
  • Contains all necessary components
Caution: For research use only.