Express Eingabe
Austria
ContactUsLinkComponent
 
ReadiUse™ TCA Deproteinization Sample Preparation Kit
Catalog # AATB19501
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
ReadiUse™ TCA Deproteinization Sample Preparation Kit
Catalog # AATB19501
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • VE:
    200 Tests
  • Bezeichnung:
    ReadiUse™ TCA deproteinization sample preparation kit

Specifications

About this item

The analysis of amino acids, ions, metabolites and other small molecules is frequently hindered by the interference of lipids, protein and enzymes present in biofluids, cell and tissue lysates.

  • Contains all necessary components
  • Useful for preparing small molecules
  • Can be used in small- and large-scale
Caution: For research use only.