ContactUsLinkComponent
Cell Navigator® Lysosome Staining Kit *Deep Red Fluorescence*
Cell Navigator® Lysosome Staining Kit *Deep Red Fluorescence*
Catalog # AATB22659
Supplier:  AAT Bioquest
CAS Number:  
undefined
Cell Navigator® Lysosome Staining Kit *Deep Red Fluorescence*
Catalog # AATB22659
Supplier:  AAT Bioquest
CAS Number:  
Specifications
  • Pk:
    500 Tests
  • Description:
    Cell Navigator™ lysosome staining kit *deep red fluorescence*
Specifications
About this item
Lysosomes are cellular organelles which contain acid hydrolase enzymes to break up waste materials and cellular debris.
  • Cell-permeant staining
  • Improved signal
  • Suitable for all major fluorescence platforms
Caution: For research use only.