Cell Explorer™ Fixable Live Cell Tracking Kit *Red Fluorescence*
Catalog # AATB22625
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pk:200 Tests
- Description:Cell Explorer™ Fixable live cell tracking kit *red fluorescence*
Specifications
About this item
Our Cell Explorer™ fluorescence imaging kits are a set of tools for labelling cells for fluorescence microscopic investigations of cellular functions.
- Long label retention
- Suitable for all major fluorescence platforms
- Contains all necessary components
- Multiple applications
Caution: For research use only.