Live or Dead™ Fixable Dead Cell Staining Kit *Deep Red Fluorescence*
Catalog # AATB22604
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pk:200 Tests
- Description:Live or Dead™ fixable dead cell staining kit *deep red fluorescence*
Specifications
About this item
Our Live or Dead™ fixable dead cell staining kits are a set of tools for labelling cells for fluorescence microscopic investigations of cell functions.
- Contains all necessary components
- Bright and stable signal
- Suitable for all major fluorescence platforms
Caution: For research use only.