Order Entry
Austria
ContactUsLinkComponent
Cell Meter™ Generic Fluorimetric Caspase Binding Assay Kit *Red Fluorescence Optimized for Flow Cytometry*
Cell Meter™ Generic Fluorimetric Caspase Binding Assay Kit *Red Fluorescence Optimized for Flow Cytometry*
Catalog # AATB22822
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Cell Meter™ Generic Fluorimetric Caspase Binding Assay Kit *Red Fluorescence Optimized for Flow Cytometry*
Catalog # AATB22822
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    100 Tests
  • Description:
    Cell Meter™ generic fluorimetric caspase binding assay kit *red fluorescence optimized for flow cytometry*
  • Environmentally Preferable:

Specifications

About this item

Our Cell Meter™ assay kits are a set of tools for monitoring cell viability.

  • Contains all necessary components
  • Multiple research applications
  • Bright and stable signal
  • Optimised for flow cytometry but suitable for all major fluorescence platforms
Caution: For research use only.