Buccutite™ Rapid PE Antibody Labelling Kits (2 Labellings)
Supplier: AAT BIOQUEST
Certificates
About this item
R-Phycoerythrin (PE) is an orange fluorescent protein which has an excitation wavelength of 565 nm and an emission wavelength of 575 nm.
- High conjugation yield
- Stable crosslinking
- Minimal hands-on time
- Robust protocol
Related Information
Manual Antibody Labelling Kits (2 Labelings).pdf Manual Buccutite Rapid Pe Antibody Labeling Kit Microscale Optimized for Labeling 100 µg Antibody per Reaction.pdf Manual Buccutite™ Rapid PE Antibody Labelling Kits.PDF Manual Buccutite™Rapid PE Antibody Labeling Kit Microscale Optimized for Labeling 25 ug Antibody Per Reaction.pdf
Manual
Frequently Bought Together