ReadiPrep™ Nuclear/Cytoplasmic Fractionation Kit (50 Tests)
Catalog # AATB60000
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pk:50 Tests
- Description:ReadiPrep™ nuclear/cytoplasmic fractionation kit
Specifications
About this item
The ReadiPrep™ Nuclear/Cytoplasmic Fractionation Kit offers stepwise isolation of nuclear and cytoplasmic extract from mammalian cells or tissue.
- Ready to use
- Multiple applications
- Compatible with most major downstream applications
Caution: For research use only.