Screen Quest™ Calbryte-520 Probenecid-Free and Wash-Free Calcium Assay Kits
Supplier: AAT Bioquest
Certificates
About this item
Calcium flux assays are the preferred methods in drug discovery for screening G protein coupled receptors (GPCR).
- 100 to 250 times fluorescence increase once bound to calcium
- No probenecid required
- Contains all necessary components
- No wash step required
- Robust and adaptable protocol
- Brighter and more photostable signal
Caution: For research use only.
Related Information
Manual Calbryte-520 Probenecid-Free and Wash-Free Calcium Assay Kits.pdf Manual Calbryte-520 Probenecid-Free, Wash-Free Calcium Assay Kits.pdf Manual Screen Quest™ Calbryte 520 Probenecid Free And Wash Free Calcium Assay Kit.pdf Manual Screen Quest™ Calbryte-520 Probenecid-Free and Wash-Free Calcium Assay Kit.pdf
Manual
Frequently Bought Together