Live or Dead™ Cell Viability Assay Kit *Red/Blue Dual Fluorescence*
AATB22788
:
- Pk:200 Tests
- Description:Live or Dead™ cell viability assay kit *red/blue dual fluorescence*
- Environmentally Preferable:
This Live or Dead™ Cell Viability Assay Kit uses two fluorescent indicators: Cellbrite™ Red (Ex/Em = 613/631 nm) for labelling viable cells and a cell-impermeable DNA-binding dye Nuclear Blue™ DCS1 (Ex/Em = 360/450 nm) for labelling dead cells with damaged membranes.
- Contains all necessary components
- Multiple research applications
- Bright and stable signal
- Suitable for all major fluorescence platforms
- Results in under 2 hours
: For research use only.