ReadiLink™ Rapid Cy5 Antibody Labeling Kit "Microscale Optimized for Labeling 50 µg Antibody Per Reaction" (2 Labelings)
Catalog # AATB1292
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pk:2 Tests
- Description:ReadiLink™ rapid Cy5 antibody labelling kit 'Microscale optimized for labelling 50 µg antibody per reaction'
Specifications
About this item
Cy5 is one of the most popular fluorescent labelling dyes for preparing orange-red fluorescent bioconjugates.
- Contains all essential components
- Minimal hands on time
- Ready to use
- Robust and simple antibody labeling protocol
- No column purification required
Caution: For research use only.